missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MLYCD Polyclonal antibody specifically detects MLYCD in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | MLYCD |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 4.1.1.9, hMCD, malonyl-CoA decarboxylase, malonyl-CoA decarboxylase, mitochondrial, MCDmalonyl coenzyme A decarboxylase, MGC59795 |
| Host Species | Rabbit |
| Immunogen | malonyl-CoA decarboxylase. (Uniprot# O95822) |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?