missing translation for 'onlineSavingsMsg'
Learn More

MLXIP Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18427246 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427246 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427246 Supplier Novus Biologicals Supplier No. NBP281965AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MLXIP Polyclonal antibody specifically detects MLXIP in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLXIP
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias bHLHe36BHLHE36, Class E basic helix-loop-helix protein 36, KIAA0867MondoA, MIRMLX-interacting protein, MLX interacting protein, MONDOAMlx interactor, Transcriptional activator MondoA
Host Species Rabbit
Immunogen MLXIP antibody was raised against a peptide corresponding to 19 amino acids near the amino terminus of human MLXIP. Amino Acid Squence: SPHREHPPKGYDFDTVNK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22877
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.