missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MLX Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148812
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The product of this gene belongs to the family of basic helix-loop-helix leucine zipper (bHLH-Zip) transcription factors. These factors form heterodimers with Mad proteins and play a role in proliferation, determination and differentiation. This gene product may act to diversify Mad family function by its restricted association with a subset of the Mad family of transcriptional repressors, namely, Mad1 and Mad4. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
Specifications
| MLX | |
| Polyclonal | |
| Unconjugated | |
| Mlx | |
| bHLHd13; BigMax alpha; BigMax protein; class D basic helix-loop-helix protein 13; hypothetical protein LOC508531; im:7146242; MAD7; MAX dimerization protein MLX; MAX-like bHLHZIP protein; max-like factor protein; max-like protein X; Mlx; MLX, MAX dimerization protein; MXD7; Protein BigMax; TCFL4; TF4; transcription factor like 4; transcription factor-like 4; transcription factor-like protein 4; wu:fb61a01; wu:fb99c01; zgc:112409 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 6945 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9UH92 | |
| Mlx | |
| E. coli-derived recombinant human MLX, Glu197-Tyr298. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction