missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLX Mouse anti-Human, Clone: PCRP-MLX-1G8, Novus Biologicals™
Mouse Monoclonal Antibody
£215.00 - £396.00
Specifications
| Antigen | MLX |
|---|---|
| Clone | PCRP-MLX-1G8 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18328366
|
Novus Biologicals
NBP3-07337-20UG |
20 μg |
£215.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18325233
|
Novus Biologicals
NBP3-07337-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MLX Monoclonal specifically detects MLX in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
| MLX | |
| 0.2 mg/ml | |
| Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| BHLHD13, bHLHd13Max-like bHLHZip protein, BigMax protein, Class D basic helix-loop-helix protein 13, MAD7, MAX-like protein X, MXD7, TCFL4Protein BigMax, transcription factor-like 4, Transcription factor-like protein 4 | |
| Recombinant full-length human MLX protein | |
| Primary | |
| Store at 4°C. |
| PCRP-MLX-1G8 | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 6945 | |
| IgG2b | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title