missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLX Antibody (PCRP-MLX-1G8), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08241H
This item is not returnable.
View return policy
Description
MLX Monoclonal antibody specifically detects MLX in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
| MLX | |
| Monoclonal | |
| HRP | |
| BHLHD13, bHLHd13Max-like bHLHZip protein, BigMax protein, Class D basic helix-loop-helix protein 13, MAD7, MAX-like protein X, MXD7, TCFL4Protein BigMax, transcription factor-like 4, Transcription factor-like protein 4 | |
| Recombinant full-length human MLX protein (Uniprot: Q9UH92) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence, Protein Array | |
| PCRP-MLX-1G8 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 6945 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction