missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MLS 1547

Product Code. 18757326
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18757326 25 mg 50mg
18749496 5 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18757326 Supplier Tocris Bioscience™ Supplier No. 6171/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

G protein-biased D2 partial agonist

MLS 1547 is a G protein-biased D2 receptor partial agonist (Ki = 1.2 μM). Stimulates D2 receptor G protein-mediated signaling and acts as an antagonist of dopamine-mediated β-arrestin recruitment at D2 receptors.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chloro-7-[[4-(2-pyridinyl)-1-piperazinyl]methyl]-8-quinolinol
CAS 315698-36-3
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C19H19ClN4O
Formula Weight Observed MW: 354.83
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.