missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MLR 1023

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective allosteric activator of Lyn kinase (EC50 = 63 nM); displays no significant activity against a range of 47 other kinases, including other Src family kinases such as Fyn, Lck and Src kinase. Reduces blood glucose levels without affecting in vivo insulin secretion. Orally bioavailable.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 202.21 |
| Purity | >99% |
| Molecular Weight (g/mol) | 202.213 |
| Chemical Name or Material | MLR 1023 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction