missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MLR 1023
GREENER_CHOICE

Product Code. 30001588
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001588

Brand: Tocris Bioscience™ 4582/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective allosteric activator of Lyn kinase

Selective allosteric activator of Lyn kinase (EC50 = 63 nM); displays no significant activity against a range of 47 other kinases, including other Src family kinases such as Fyn, Lck and Src kinase. Reduces blood glucose levels without affecting in vivo insulin secretion. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41964-07-2
Molecular Formula C11H10N2O2

Specifications

Quantity 10 mg
Formula Weight 202.21
Purity >99%
Molecular Weight (g/mol) 202.213
Chemical Name or Material MLR 1023
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.