missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLN64 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20292-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of a subfamily of lipid trafficking proteins that are characterized by a C-terminal steroidogenic acute regulatory domain and an N-terminal metastatic lymph node 64 domain. The encoded protein localizes to the membranes of late endosomes and may be involved in exporting cholesterol. Alternative splicing results in multiple transcript variants.Specifications
| MLN64 | |
| Polyclonal | |
| Unconjugated | |
| STARD3 | |
| CAB1, MLN 64, MLN64, Protein CAB1, STARD3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10948, 363675, 59045 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14849, Q61542 | |
| Stard3 | |
| MLN64 Fusion Protein Ag14061 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction