missing translation for 'onlineSavingsMsg'
Learn More

MLN2480, TRC

Product Code. 30480218
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480218 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30480218 Supplier Toronto Research Chemicals Supplier No. TRCM4251401MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material MLN2480
CAS 1096708-71-2
Molecular Formula C17H12Cl2F3N7O2S
Quantity 1 mg
Synonym 6-Amino-5-chloro-N-[(1R)-1-[5-[[[5-chloro-4-(trifluoromethyl)-2-pyridinyl]amino]carbonyl]-2-thiazolyl]ethyl]-4-pyrimidinecarboxamide, (R)-2-(1-(6-Amino-5-chloropyrimidine-4-carboxamido)ethyl)-N-(5-chloro-4-(trifluoromethyl)pyridin-2-yl)thiazole-5-carboxamide
InChI Formula InChI=1S/C17H12Cl2F3N7O2S/c1-6(28-15(31)12-11(19)13(23)27-5-26-12)16-25-4-9(32-16)14(30)29-10-2-7(17(20,21)22)8(18)3-24-10/h2-6H,1H3,(H,28,31)(H2,23,26,27)(H,24,29,30)/t6-/m1/s1
SMILES [Br-].CN(C)CCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC Name 2-[(1R)-1-[(6-amino-5-chloropyrimidine-4-carbonyl)amino]ethyl]-N-[5-chloro-4-(trifluoromethyl)pyridin-2-yl]-1,3-thiazole-5-carboxamide
Molecular Weight (g/mol) 506.29
Formula Weight 505.0102
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.