missing translation for 'onlineSavingsMsg'
Learn More

MLN0905, MedChemExpress

Product Code. p-200098552
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. Quantity unitSize
16496328 10 mg 10mg
16458917 5 mg 5mg
16432447 50 mg 50mg
3 options
This item is not returnable. View return policy

Product Code. 16496328

Brand: Medchem Express HY15155

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress MLN0905 is a potent PLK1 inhibitor, with an IC50 of 2 nM.

MLN0905 is a potent PLK1 inhibitor, with an IC50 of 2 nM.

Specifikationer

Chemical Name or Material MLN0905
CAS 1228960-69-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H25F3N6S
Quantity 10 mg
Synonym PLK1 Inhibitor
Solubility Information DMSO : ≥ 30 mg/mL (61.66 mM)
SMILES CC1=NC=C(CCCN(C)C)C=C1NC2=NC(C(C=CC(C(F)(F)F)=C3)=C3NC(C4)=S)=C4C=N2
Molecular Weight (g/mol) 486.56
Formula Weight 486.56
Percent Purity 95.32%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Visa mer Visa mindre

Research purposes only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.