missing translation for 'onlineSavingsMsg'
Learn More

MLLT6 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18421224 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421224 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421224 Supplier Novus Biologicals Supplier No. NBP122995R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MLLT6 Polyclonal antibody specifically detects MLLT6 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLLT6
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias AF17Myeloid/lymphoid or mixed-lineage leukemia, translocated to, 6, ALL1-fused gene from chromosome 17 protein, FLJ23480, myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 6, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocatedto, 6, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 6, protein AF-17, trithorax homolog
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1043 and 1093 of human AF17 (MLLT6, myeloid/lymphoid or mixed-lineage leukemia translocated to, 6 ) using the numbering given in entry NP_005928.2 (GeneID 4302).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4302
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.