missing translation for 'onlineSavingsMsg'
Learn More

MLLT6 Antibody (1B2), Novus Biologicals™

Product Code. 18364638 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364638 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364638 Supplier Novus Biologicals Supplier No. H00004302M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MLLT6 Monoclonal antibody specifically detects MLLT6 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLLT6
Applications Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA
Classification Monoclonal
Clone 1B2
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH07237
Gene Alias AF17Myeloid/lymphoid or mixed-lineage leukemia, translocated to, 6, ALL1-fused gene from chromosome 17 protein, FLJ23480, myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog),translocated to, 6, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog), translocatedto, 6, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila),translocated to, 6, protein AF-17, trithorax homolog
Host Species Mouse
Immunogen MLLT6 (AAH07237, 1 a.a. ~ 462 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGAVNPLLSQAESSHTEPDLEDCSFRCRGTSPQESLSSMSPISSLPALFDQTASAPCGGGQLDPAAPGTTNMEQLLEKQGDGEAGVNIVEMLKALHALQKENQRLQEQILSLTAKKERLQILNVQLSVPFPALPAALPAANGPVPGPYGLPPQAGSSDSLSTSKSPPGKSSLGLDNSLSTSSEDPHSGCPSRSSSSLSFHSTPPPLPLLQQSPATLPLALPGAPAPLPPQPQNGLGRAPGAAGLGAMPMAEGLLGGLAGSGGLPLNGLLGGLNGAAAPNPASLSQAGGAPTLQLPGCLNSLTEQQRHLLQQQEQQLQQLQQLLASPQLTPEHQTVVYQMIQQIQQKRELQRLQMAGGSQLPMASLLAGSSTPLLSAGTPGLLPTASAPPLLPAGALVAPSLGNNTSLMAAAAAAAAVAAAGGPPVLTAQTNPFLSLSGAEGSGGGPKGGTADKGASANQEKG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4302
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.