missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLLT11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA551184
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total MLLT11 / AF1Q protein.
The gene variously symbolized ALL1, HRX, or MLL located on 11q23 has been demonstrated to be fused with a number of translocation partners in cases of leukemia. t(1;11)(q21;q23) translocations that fused the MLL gene to a gene on chromosomal band 1q21 in 2 infants with acute myelomonocytic leukemia have been demonstrated. The N-terminal portion of the MLL gene is critical for leukemogenesis in translocations involving band 11q23. This gene encodes 90 amino acids. It was found to be highly expressed in the thymus but not in peripheral lymphoid tissues. In contrast to its restricted distribution in normal hematopoietic tissue, this gene was expressed in all leukemic cell lines tested.Specifications
MLLT11 | |
Polyclonal | |
Unconjugated | |
MLLT11 | |
AF1Q; AF1Q protein; AI839562; ALL1 fused gene from chromosome 1q; ALL1-fused gene from chromosome 1q; MLLT11; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 11; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 11; protein AF1q; RGD1305525; Zfp692; zinc finger protein 692 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10962 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q13015 | |
MLLT11 | |
Full length fusion protein of human MLLT11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |