missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MLLT10 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA519296
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, human and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Translocations affecting chromosome 11q23 involve many partner chromosome regions and occur in various leukemias. The 11q23 gene involved in the translocations is MLL. MLLT10 is the partner gene to MLL1 involved in t(10;11)(p12;q23) translocations. In an analysis of two leukemia patients, the in t(10;11)(p12;q23) translocation fuses MLL1, a SET domain containg histone methyltransferase, to the MLLT10 gene. The MLLT10 gene encodes a predicted 1,027-amino acid protein containing an N-terminal zinc finger and a C-terminal leucine zipper domain. The MLLT10 gene is one of the few MLL partner genes to be independently rearranged with a third gene in leukemia, the CALM gene in the t(10;11)(p12;q14) translocation. Chimeric fusion proteins MLL/AF10 and CALM/AF10 consistently retain the leucine zipper motif of MLLT10. The leucine zipper interacts with GAS41, a protein previously identified as the product of an amplified gene in a glioblastoma. GAS41 interacts with integrase interactor-1 (INI1), a component of the SWI/SNF complex, which acts to remodel chromatin and to modulate transcription. Retention of the leucine zipper in the MLL and CALM fusions suggested that a key feature of these chimeric proteins may be their ability to interfere in normal gene regulation through interaction with the adenosine triphosphate-dependent chromatin remodeling complexes.
Specifications
| MLLT10 | |
| Polyclonal | |
| Unconjugated | |
| MLLT10 | |
| Af10; af-10; ALL1-fused gene from chromosome 10 protein; B130021D15Rik; D630001B22Rik; DKFZp686E10210; MGC75086; mKIAA4140; MLLT10; myeloid/lymphoid or mixed lineage-leukemia translocation to 10 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 10; myeloid/lymphoid or mixed-lineage leukemia translocated to, 10; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 10; protein AF-10; translocated to, 10; type I AF10 protein; type III AF10 protein; type IV AF10 protein | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 361285, 8028 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P55197 | |
| MLLT10 | |
| Synthetic peptide sequence (RLEDTTARFTNAN) corresponding to the internal amino acids of MLLT10 (aa 281-293). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction