missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLLT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35824-100ul
This item is not returnable.
View return policy
Description
MLLT1 Polyclonal antibody specifically detects MLLT1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| MLLT1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| ENLENL/mLL fusion, LTG19MLLT1/mLL fusion, myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 1, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 1, protein ENL, YEATS domain-containing protein 1, YEATS1myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocatedto, 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 190-270 of human MLLT1 (NP_005925.2).,, Sequence:, KTSKPHKVTKEHRERPRKDSESKSSSKELEREQAKSSKDTSRKLGEGRLPKEEKAPPPKAAFKEPKMALKETKLESTSPKG | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4298 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto