missing translation for 'onlineSavingsMsg'
Learn More

MLLT1 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18417577 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417577 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417577 Supplier Novus Biologicals Supplier No. NBP126652AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MLLT1 Polyclonal antibody specifically detects MLLT1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLLT1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ENLENL/MLL fusion, LTG19MLLT1/MLL fusion, myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 1, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 1, protein ENL, YEATS domain-containing protein 1, YEATS1myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocatedto, 1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 150 and 200 of human myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 1 using the numbering given in entry NP_005925.2 (GeneID 4298).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4298
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.