missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MLL5 Polyclonal antibody specifically detects MLL5 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | MLL5 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | PE |
| Formulation | PBS |
| Gene Alias | EC 2.1.1.43, FLJ10078, HDCMC04P, histone-lysine N-methyltransferase MLL5, KMT2EFLJ14026, Lysine N-methyltransferase 2E, MGC70452, myeloid/lymphoid or mixed-lineage leukemia 5 (trithorax homolog, Drosophila), Myeloid/lymphoid or mixed-lineage leukemia protein 5 |
| Host Species | Rabbit |
| Immunogen | myeloid/lymphoid or mixed-lineage leukemia 5(trithorax homolog, Drosophila). (Uniprot# Q8IZD2) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?