missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Research Category | Cancer, Cell Biology, Cell Cycle and Replication, Stem Cells |
| Target | MLL5 |
| Synonym | EC 2.1.1.43, FLJ10078, HDCMC04P, histone-lysine N-methyltransferase MLL5, KMT2EFLJ14026, Lysine N-methyltransferase 2E, MGC70452, myeloid/lymphoid or mixed-lineage leukemia 5 (trithorax homolog, Drosophila), Myeloid/lymphoid or mixed-lineage leukemia protein 5 |
| Gene Symbol | MLL5 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?