missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MLL3 Antibody (7D5D7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27146-0.1ml
This item is not returnable.
View return policy
Description
MLL3 Monoclonal antibody specifically detects MLL3 in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot
Specifications
| MLL3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| ALR-like protein, DKFZp686C08112, EC 2.1.1.43, FLJ12625, HALRMGC119851, histone-lysine N-methyltransferase MLL3, histone-lysine N-methyltransferase, H3 lysine-4 specific, Homologous to ALR protein, KIAA1506FLJ38309, KMT2C, KMT2CMGC119852, Lysine N-methyltransferase 2C, MGC119853, myeloid/lymphoid or mixed-lineage leukemia 3, Myeloid/lymphoid or mixed-lineage leukemia protein 3 | |
| Purified recombinant fragment of human MLL3 (AA: 1-205) expressed in E. Coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Western Blot | |
| 7D5D7 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 58508 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction