missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MLKL Recombinant Rabbit Monoclonal Antibody (24GB4620)

Rabbit Recombinant Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | MLKL |
|---|---|
| Clone | 24GB4620 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275748
![]() |
Invitrogen™
MA557484 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276220
|
Invitrogen™
MA557485 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MLKL Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.Specifications
| MLKL | |
| 1 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8NB16, Q9D2Y4 | |
| 197259, 74568 | |
| A synthesized peptide derived from human MLKL (aa 371-472). | |
| Antibody |
| 24GB4620 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| Mlkl | |
| 9130019I15Rik; hMLKL; mixed lineage kinase domain like pseudokinase; mixed lineage kinase domain-like; mixed lineage kinase domain-like protein; Mlkl; rCG51567-like | |
| Mlkl | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title