missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLKL Rabbit anti-Human, Mouse, Rat, Clone: 7L1P5, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £386.00
Specifications
| Antigen | MLKL |
|---|---|
| Clone | 7L1P5 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18379203
|
Bio-Techne
NBP3-15859-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18369125
|
Novus Biologicals
NBP3-15859-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MLKL Monoclonal antibody specifically detects MLKL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| MLKL | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 10-193 of human MLKL (Q8NB16). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 7L1P5 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Necroptosis, Protein Kinase | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 197259 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title