missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLK4 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-41080PE
This item is not returnable.
View return policy
Description
MLK4 Polyclonal antibody specifically detects MLK4 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)
Specifications
| MLK4 | |
| Polyclonal | |
| Immunoprecipitation, Western Blot (Negative) | |
| dJ862P8.3, EC 2.7.11, EC 2.7.11.25, mitogen-activated protein kinase kinase kinase MLK4, mixed lineage kinase 4, MLK4, RP5-862P8.2 | |
| The immunogen for this product maps to a region between residue 650 and 700 of human mixed lineage kinase 4 using the numbering given in entry NP_115811.2 (GeneID 84451). | |
| 0.1 mL | |
| Protein Kinase | |
| 84451 | |
| Store at 4C in the dark. | |
| IgG |
| Immunoprecipitation | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction