missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MLK3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544553
This item is not returnable.
View return policy
Description
Peptide sequence: PVGQRSAKSP RREEEPRGGT VSPPPGTSRS APGTPGTPRS PPLGLISRPR Sequence homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
MAP3K11 (MLK3) is a serine/threonine kinase that activates both the p38 MAP kinase and c-Jun kinase pathways. This kinase contains a SH3 domain and a leucine zipper-basic motif. This kinase preferentially activates MAPK8/JNK kinase, and functions as a positive regulator of JNK signaling pathway. This kinase can directly phosphorylate, and activates IkappaB kinase alpha and beta, and is found to be involved in the transcription activity of NF-kappaB mediated by Rho family GTPases and CDC42.
Specifications
| MLK3 | |
| Polyclonal | |
| Unconjugated | |
| MAP3K11 | |
| 2610017K16Rik; MAP3K11; MEKK11; MGC17114; mitogen activated protein kinase kinase kinase 11; mitogen-activated protein kinase kinase kinase 11; Mixed lineage kinase 3; Mlk3; MLK-3; protein-tyrosine kinase PTK1; PTK1; RHOE; SH3 domain-containing proline-rich kinase; SPRK; Src-homology 3 domain-containing proline-rich kinase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4296 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q16584 | |
| MAP3K11 | |
| Synthetic peptide directed towards the middle region of human MAP3K11. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction