missing translation for 'onlineSavingsMsg'
Learn More

MLK2 Antibody, Novus Biologicals™

Product Code. 18394052 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394052 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394052 Supplier Novus Biologicals Supplier No. NBP312199

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MLK2 Polyclonal antibody specifically detects MLK2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLK2
Applications Western Blot, ELISA, Immunoprecipitation, Dot Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:4000, Immunoprecipitation 1:150, Dot Blot 1:4000
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias EC 2.7.11, MEKK10, mitogen-activated protein kinase kinase kinase 10, Mixed lineage kinase 2, MKN28 kinase, MLK2MKN28 derived nonreceptor_type serine/threonine kinase, MSTEC 2.7.11.25, Protein kinase MST
Host Species Rabbit
Immunogen Synthetic peptide correpsonding to the C-terminal taken within amino acid region 900-950 on human MAP3K10 protein.
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 4294
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.