missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MLi-2
GREENER_CHOICE

Product Code. 18799746
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18799746

Brand: Tocris Bioscience™ 5756/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective LRRK2 inhibitor

MLi-2 is a potent and selective LRRK2 inhibitor (IC50 = 0.76-3.4 nM depending on assay), which exhibits >295-fold selectivity for over 300 kinases and a diverse panel of receptors and ion channels. MLi-2 is centrally bioavailable and active in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rel-3-[6-[(2R,6S)-2,6-Dimethyl-4-morpholinyl]-4-pyrimidyl]-5-[(1-methylcyclopropyl)oxy]-1H-indazole
CAS 1627091-47-7
Quantity 50 mg
Solubility Soluble to 50mM in DMSO
Molecular Formula C21H25N5O2
Formula Weight 379.46
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.