missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLH3 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£145.00 - £351.00
Specifications
| Antigen | MLH3 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232986
|
Novus Biologicals
NBP3-33483-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229779
|
Novus Biologicals
NBP3-33483-100ul |
100 μL |
£351.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MLH3 Monoclonal antibody specifically detects MLH3 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| MLH3 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, Mismatch Repair | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 27030 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DNA mismatch repair protein Mlh3, HNPCC7, MGC138372, mutL (E. coli) homolog 3, mutL homolog 3 (E. coli), MutL protein homolog 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1190-1389 of human MLH3 (NP_001035197.1).,, Sequence:, QVLQQVDNKFIACLMSTKTEENGEAGGNLLVLVDQHAAHERIRLEQLIIDSYEKQQAQGSGRKKLLSSTLIPPLEITVTEEQRRLLWCYHKNLEDLGLEFVFPDTSDSLVLVGKVPLCFVEREANELRRGRSTVTKSIVEEFIREQLELLQTTGGIQGTLPLTVQKVLASQACHGAIKFNDGLSLQESCRLIEALSSCQL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title