missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLH1 Mouse anti-Human, Clone: G168-728, Novus Biologicals™
Mouse Monoclonal Antibody
£388.00
Specifications
| Antigen | MLH1 |
|---|---|
| Clone | G168-728 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 2 to 4 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
MLH1 Monoclonal specifically detects MLH1 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| MLH1 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mismatch Repair, Tumor Suppressors | |
| 10 mM PBS with 0.05% BSA | |
| 4292 | |
| IgG1 κ | |
| Protein A or G purified |
| G168-728 | |
| Immunohistochemistry-Paraffin 2 to 4 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| COCA2FCC2, DNA mismatch repair protein Mlh1, hMLH1, HNPCC, HNPCC2MGC5172, mutL (E. coli) homolog 1 (colon cancer, nonpolyposis type 2), mutL homolog 1, colon cancer, nonpolyposis type 2 (E. coli), MutL protein homolog 1 | |
| Recombinant full-length human MLH1 protein | |
| Primary | |
| Store at 4°C. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto