missing translation for 'onlineSavingsMsg'
Learn More

MLH1 Antibody (rMLH1/6285), Novus Biologicals™

Mouse Monoclonal Antibody

£195.00 - £385.00

Specifications

Antigen MLH1
Clone rMLH1/6285
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18631695
View Documents
Novus Biologicals
NBP3-20610-100ug
100 μg
£385.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18669743
View Documents
Novus Biologicals
NBP3-20610-20ug
20 μg
£195.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MLH1 Monoclonal antibody specifically detects MLH1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications

Specifications

MLH1
0.2 mg/ml
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mismatch Repair, Tumor Suppressors
10mM PBS with 0.05% BSA
4292
IgG1 κ
Protein A or G purified
rMLH1/6285
Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL
Monoclonal
Purified
RUO
Human
COCA2FCC2, DNA mismatch repair protein Mlh1, hMLH1, HNPCC, HNPCC2MGC5172, mutL (E. coli) homolog 1 (colon cancer, nonpolyposis type 2), mutL homolog 1, colon cancer, nonpolyposis type 2 (E. coli), MutL protein homolog 1
Recombinant full-length human MLH1 protein
Primary
Store at 4&drg;C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.