missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11835-1-AP-20UL
This item is not returnable.
View return policy
Description
MLF2 is a protein coding gene.Specifications
| MLF2 | |
| Polyclonal | |
| Unconjugated | |
| MLF2 | |
| MLF2, myeloid leukemia factor 2, NTN4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 30853, 312709, 8079 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15773, Q99KX1 | |
| Mlf2 | |
| MLF2 Fusion Protein Ag2270 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction