missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MLF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552754
This item is not returnable.
View return policy
Description
Immunogen sequence: MIGNMEHMTA GGNCQTFSSS TVISYSNTGD GAPKVYQETS EMRSAPGGIR ETRRTVRDSD SGLEQMSIGH HIRDRAHILQ RSRNHRTGDQ EERQDYINLD ESEAAAFDDE WRRETSRFRQ QRPLEFRRLE SSGA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
MLF2 is a protein coding gene.
Specifications
| MLF2 | |
| Polyclonal | |
| Unconjugated | |
| MLF2 | |
| AI256856; AU018944; C77284; MLF2; Myelodysplasia-myeloid leukemia factor 2; myeloid leukemia factor 2; myeloid leukemia factor 2 homolog; NTN4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8079 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15773 | |
| MLF2 | |
| Recombinant protein corresponding to Human MLF2. Recombinant protein control fragment (Product #RP-90014). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction