Learn More
Invitrogen™ MLF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140305
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: NQEFINMNES DAHAFDEEWQ SEVLKYKPGR HNLGNTRMRS VGHENPGSRE LKRREKPQQS PAIEHGRRSN VLGDKLHIKG SSVKSNK Highest antigen sequence indentity to the following orthologs: Mouse - 75%, Rat - 68%.
SERBP1 may play a role in the regulation of mRNA stability. It binds to the 3'-most 134 nt of the SERPINE1/PAI1 mRNA, a region which confers cyclic nucleotide regulation of message decay.
Specifications
| MLF1 | |
| Polyclonal | |
| Unconjugated | |
| MLF1 | |
| Hematopoietic lineage switch 7; HLS7; Mlf1; MLF-1; Myelodysplasia-myeloid leukemia factor 1; myeloid leukemia factor 1; testis tissue sperm-binding protein Li 49e; Unknown (protein for MGC:133499) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4291 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P58340 | |
| MLF1 | |
| Recombinant protein corresponding to Human MLF1. Recombinant protein control fragment (Product #RP-109698). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.