missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MLF1 Monoclonal Antibody (OTI3A3)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525975
This item is not returnable.
View return policy
Description
MLF1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
SERBP1 may play a role in the regulation of mRNA stability. It binds to the 3'-most 134 nt of the SERPINE1/PAI1 mRNA, a region which confers cyclic nucleotide regulation of message decay.
Specifications
| MLF1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P58340 | |
| MLF1 | |
| Full length human recombinant protein of MLF1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI3A3 | |
| Unconjugated | |
| MLF1 | |
| Hematopoietic lineage switch 7; HLS7; Mlf1; MLF-1; Myelodysplasia-myeloid leukemia factor 1; myeloid leukemia factor 1; testis tissue sperm-binding protein Li 49e; Unknown (protein for MGC:133499) | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4291 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction