missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLF1 Interacting Protein, p Thr78 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77938
This item is not returnable.
View return policy
Description
MLF1 Interacting Protein Polyclonal specifically detects MLF1 Interacting Protein in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| MLF1 Interacting Protein (p Thr78) | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| CENP-50, CENP-U, CENPU50, CENPUMLF1-interacting protein, centromere protein of 50 kDa, centromere protein U, FLJ23468, ICEN24, Interphase centromere complex protein 24, KLIP1CENP50, KSHV latent nuclear antigen interacting protein 1, KSHV latent nuclear antigen-interacting protein 1, MLF1 interacting protein, PBIP1, Polo-box-interacting protein 1 | |
| Rabbit | |
| 47.52 kDa | |
| 0.1 mg | |
| Phospho Specific | |
| 79682 | |
| Human, Bovine, Canine, Primate | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot | |
| 1 mg/ml | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:25000, Immunohistochemistry 20 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Dot Blot | |
| NP_078905 | |
| CENPU | |
| This affinity purified MLF1 Interacting Protein [p Thr78] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids surrounding Thr78 of human MLF1 Interacting Protein protein. The immunogen peptide is phosphorylated at Thr78. (Uniprot: Q71F23) | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is specific for human MLF1 Interacting Protein protein phosphorylated at Thr78. | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction