missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MLCK (MLCK2), GST Tag Recombinant Protein

Product Code. 11573567
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11573567

Brand: Invitrogen™ PV3835

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length.

MLCK (MLCK2), now also known as MYLK3, is of the myosin regulatory light chain kinases which facilitates muscle contraction. MLCK, a member of the Ser/Thr protein kinase family, is responsible for smooth muscle contraction via phosphorylation of a specific serine in the N-terminus of myosin light chains (MLC), an event that facilitates myosin interaction with actin filaments. It is a central determinant in the development of vascular permeability and tissue edema formation. In the nervous system it has been shown to control the growth initiation of astrocytic processes in culture and to participate in transmitter release at synapses formed between cultured sympathetic ganglion cells. MLCK acts as a critical participant in signaling sequences that result in fibroblast apoptosis. Smooth muscle and non-muscle isozymes are expressed in a wide variety of adult and fetal tissues and in cultured endothelium with qualitative expression appearing to be neither tissue- nor development-specific. Non-muscle isoform 2 is the dominant splice variant expressed in various tissues. The Telokin isoform, which binds calmodulin, has been found in a wide variety of adult and fetal tissues. MLCK is probably down-regulated by phosphorylation. The protein contains 1 fibronectin type III domain and 9 immunoglobulin-like C2-type domains.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q32MK0
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.5mM EDTA, 2mM DTT, 50% glycerol, 150mM NaCl, 0.02% Triton X-100 and no preservative; pH 7.5
Gene ID (Entrez) 91807
Molecular Weight (g/mol) 113.6 kDa
Name Human MLCK (MLCK2), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias caMLCK; cardiac MLCK; cardiac-MLCK; cardiac-MyBP-C associated Ca/CaM kinase; cardiac-MyBP-C-associated Ca/CaM kinase; cardiac-specific myosin light chain kinase; D830007F02Rik; MLC kinase; MLCK; MLCK2; Mylk3; Myosin light chain kinase 3; putative myosin light chain kinase 3; putative myosin light chain kinase 3; myosin light chain kinase 3; RGD1305801
Common Name MLCK
Gene Symbol MYLK3
KinaseFamily MLCK Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical binding/kinetic
Kinase Group Ser⁄Thr Kinases: CAMK Group
Shipping Condition Dry Ice
Tag Position N-Terminal
Target Entry MLCK
Target Gene myosin light chain kinase
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Drug Discovery: Immunology, Signal Transduction
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.