missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress MLCK inhibitor peptide 18

Product Code. 16409508
Encompass_Preferred
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 16409508

Brand: MedChemExpress HYP1029

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MLCK inhibitor peptide 18 is a myosin light chain kinase (MLCK) inhibitor with an IC50 of 50 nM, and inhibits CaM kinase II only at 4000-fold higher concentrations.

Specifications

Purity 99.66%
CAS 224579-74-2
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 1324.62 g/mol
Product Type Reference compound
Quantity 10 mg
Shipping Condition Room temperature
Solubility H2O : 100 mg/mL (75.49 mM∣Need ultrasonic)
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.