missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLC1SA Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £375.00
Specifications
| Antigen | MLC1SA |
|---|---|
| Applications | Western Blot, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18384824
|
Novus Biologicals
NBP3-03351-20UL |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18321744
|
Novus Biologicals
NBP3-03351-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MLC1SA Polyclonal specifically detects MLC1SA in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.Specifications
| MLC1SA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 140465 | |
| IgG | |
| Affinity purified |
| Western Blot, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| MLC1sa, MLC1SAmyosin light chain 1, slow-twitch muscle A isoform, myosin alkali light chain 1 slow a, myosin light chain 1 slow a, Myosin light chain 1 slow-twitch muscle A isoform, myosin light chain 6B, myosin, light chain 6B, alkali, smooth muscle and non-muscle, myosin, light polypeptide 6B, alkali, smooth muscle and non-muscle, smooth muscle and non-muscle myosin alkali light chain 6B, Smooth muscle and nonmuscle myosin light chain alkali 6B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-208 of human MLC1SA (NP_002466.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title