missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLC1SA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37928-100ul
This item is not returnable.
View return policy
Description
MLC1SA Polyclonal antibody specifically detects MLC1SA in Human samples. It is validated for ELISA,Western Blot
Specifications
| MLC1SA | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| MLC1sa, MLC1SAmyosin light chain 1, slow-twitch muscle A isoform, myosin alkali light chain 1 slow a, myosin light chain 1 slow a, Myosin light chain 1 slow-twitch muscle A isoform, myosin light chain 6B, myosin, light chain 6B, alkali, smooth muscle and non-muscle, myosin, light polypeptide 6B, alkali, smooth muscle and non-muscle, smooth muscle and non-muscle myosin alkali light chain 6B, Smooth muscle and nonmuscle myosin light chain alkali 6B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-208 of human MLC1SA (NP_002466.1).,, Sequence:, MPPKKDVPVKKPAGPSISKPAAKPAAAGAPPAKTKAEPAVPQAPQKTQEPPVDLSKVVIEFNKDQLEEFKEAFELFDRVGDGKILYSQCGDVMRALGQNPTNAEVLKVLGNPKSDELKSRRVDFETFLPMLQAVAKNRGQGTYEDYLEGFRVFDKEGNGKVMGAELRHVLTTLGEKMTEEEVETVLAGHEDSNGCINYEAFLKHILSV | |
| 100 μL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 140465 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction