missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLC1SA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | LOC100359980 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
MLC1SA Polyclonal specifically detects MLC1SA in Rat samples. It is validated for Western Blot.Specifications
| LOC100359980 | |
| Polyclonal | |
| Rabbit | |
| D3ZHA7 | |
| 100359980 | |
| IgG | |
| 23 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| smooth muscle and non-muscle myosin alkali light chain 6B-like | |
| LOC100359980 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title