missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies MLA™ Silver Brand Precision Pipetters

Click to view available options
Volume (Metric):
5 μL
15 μL
30 μL
40 μL
60 μL
70 μL
75 μL
80 μL
90 μL
120 μL
400 μL
450 μL
600 μL
700 μL
800 μL
900 μL
Unit Size:
Each
Description
- 'To deliver' capability ensures volume accuracy over a wide range of viscosities
- Ideal for repetitive delivery of reagents when high precision and accuracy are required
- Capacity engraved on bonnet
Specifications
Specifications
| Accuracy | ±5% |
| Autoclavable | Not Autoclavable |
| Color | Silver |
| Disposable | Reusable |
| For Use With | Micropipette Tip |
| Material | Aluminum Alloy, Stainless Steel |
| Model | MLA Precision |
| Number of Channels | 1 |
| Sterility | Non-sterile |
| Type | Precision Pipetter |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction