missing translation for 'onlineSavingsMsg'
Learn More

ML390, MedChemExpress

Product Code. 16493118
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493118 10 mg 10mg
16459676 50 mg 50mg
16434416 100 mg 100mg
16415857 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493118 Supplier Medchem Express Supplier No. HY100688
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ML390 is a potent dihydroorotate dehydrogenase (DHODH) inhibitor. ML390 is an inducer of myeloid differentiation and causes myeloid differentiation in murine (ER-HoxA9) and human (U937 and THP1) acute myeloid leukemia (AML) models.

ML390 is a potent dihydroorotate dehydrogenase (DHODH) inhibitor. ML390 is an inducer of myeloid differentiation and causes myeloid differentiation in murine (ER-HoxA9) and human (U937 and THP1) acute myeloid leukemia (AML) models.

Specifications

Chemical Name or Material ML390
CAS 2029049-79-2
Color White
Purity Grade Notes Research
Molecular Formula C21H21F3N2O3
Quantity 10 mg
Solubility Information DMSO : ≥ 27 mg/mL (66.44 mM)
SMILES O=C(NCCC(N[C@H]1C(C=CC=C2)=C2CCC1)=O)C3=CC=C(OC(F)(F)F)C=C3
Molecular Weight (g/mol) 406.4
Formula Weight 406.4
Percent Purity 99.96%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.