missing translation for 'onlineSavingsMsg'
Learn More

ML348, MedChemExpress

Product Code. 16459696
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16459696 50 mg 50mg
16455877 5 mg 5mg
16444436 100 mg 100mg
16413148 10 mg 10mg
16402646 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16459696 Supplier Medchem Express Supplier No. HY100736
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ML348 (GNF-Pf-1127) is a selective and reversible acyl-protein thioesterase 1 (APT1)/lysophospholipase 1 (LYPLA1) inhibitor with an IC50 of 210 nM, and barely inhibits LYPLA2.

ML348 (GNF-Pf-1127) is a selective and reversible acyl-protein thioesterase 1 (APT1)/lysophospholipase 1 (LYPLA1) inhibitor with an IC50 of 210 nM, and barely inhibits LYPLA2.

Specifications

Chemical Name or Material ML348
CAS 899713-86-1
Color White
Purity Grade Notes Research
Molecular Formula C18H17ClF3N3O3
Quantity 50 mg
Synonym GNF-Pf-1127
Solubility Information DMSO : 25 mg/mL (60.13 mM; Need ultrasonic)
SMILES O=C(NC1=CC(C(F)(F)F)=CC=C1Cl)CN2CCN(C(C3=CC=CO3)=O)CC2
Molecular Weight (g/mol) 415.79
Formula Weight 415.79
Percent Purity 99.96%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.