missing translation for 'onlineSavingsMsg'
Learn More

ML347, MedChemExpress

Product Code. p-200101908
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485048

Brand: Medchem Express HY12274

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML347 (LDN193719) is a highly selective ALK1/ALK2 inhibitor. ML347 has IC50 values of 46 and 32 nM against ALK1 and ALK2, respectively, >300-fold selective over ALK3. ML347 block the phosphorylation of Smad1/5 by TGF-β1.

ML347 (LDN193719) is a highly selective ALK1/ALK2 inhibitor. ML347 has IC50 values of 46 and 32 nM against ALK1 and ALK2, respectively, >300-fold selective over ALK3. ML347 block the phosphorylation of Smad1/5 by TGF-β1.

Specifications

Chemical Name or Material ML347
CAS 1062368-49-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H16N4O
Quantity 10 mg
Synonym LDN 193719
Solubility Information DMSO : 10 mg/mL (28.38 mM; Need ultrasonic)
SMILES COC1=CC=C(C2=CN3C(N=C2)=C(C4=C5C=CC=NC5=CC=C4)C=N3)C=C1
Molecular Weight (g/mol) 352.39
Formula Weight 352.39
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.