missing translation for 'onlineSavingsMsg'
Learn More

ML347, MedChemExpress

Product Code. 16485048
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485048 10 mg 10mg
16461287 50 mg 50mg
16427777 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16485048 Supplier Medchem Express Supplier No. HY12274

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML347 (LDN193719) is a highly selective ALK1/ALK2 inhibitor. ML347 has IC50 values of 46 and 32 nM against ALK1 and ALK2, respectively, >300-fold selective over ALK3. ML347 block the phosphorylation of Smad1/5 by TGF-β1.

ML347 (LDN193719) is a highly selective ALK1/ALK2 inhibitor. ML347 has IC50 values of 46 and 32 nM against ALK1 and ALK2, respectively, >300-fold selective over ALK3. ML347 block the phosphorylation of Smad1/5 by TGF-β1.

Specifications

Chemical Name or Material ML347
CAS 1062368-49-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H16N4O
Quantity 10 mg
Synonym LDN 193719
Solubility Information DMSO : 10 mg/mL (28.38 mM; Need ultrasonic)
SMILES COC1=CC=C(C2=CN3C(N=C2)=C(C4=C5C=CC=NC5=CC=C4)C=N3)C=C1
Molecular Weight (g/mol) 352.39
Formula Weight 352.39
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.