missing translation for 'onlineSavingsMsg'
Learn More

ML335, MedChemExpress

Product Code. 16490277
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490277 50 mg 50mg
16486637 5 mg 5mg
16484946 100 mg 100mg
16483918 10 mg 10mg
16482906 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16490277 Supplier Medchem Express Supplier No. HY104005
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ML335 is a selective activator of both TREK-1 and TREK-2.

ML335 is a selective activator of both TREK-1 and TREK-2.

Specifications

Chemical Name or Material ML335
CAS 825658-06-8
Color White
Purity Grade Notes Research
Molecular Formula C15H14Cl2N2O3S
Quantity 50 mg
Solubility Information DMSO : 155 mg/mL (415.27 mM; Need ultrasonic and warming)
SMILES O=C(NCC1=CC=C(Cl)C=C1Cl)C2=CC=C(NS(=O)(C)=O)C=C2
Molecular Weight (g/mol) 373.25
Formula Weight 373.25
Percent Purity 99.93%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.