missing translation for 'onlineSavingsMsg'
Learn More

ML324, TRC

Product Code. 30482929
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482929 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482929 Supplier Toronto Research Chemicals Supplier No. TRCM4253451MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[3-(Dimethylamino)propyl]-4-(8-hydroxy-6-quinolinyl)-benzamide
CAS 1222800-79-4
Molecular Formula C21H23N3O2
Quantity 1 mg
Synonym NCGC00183808-01, N-[3-(Dimethylamino)propyl]-4-(8-hydroxy-6-quinolinyl)-benzamide
InChI Formula InChI=1S/C21H23N3O2/c1-24(2)12-4-11-23-21(26)16-8-6-15(7-9-16)18-13-17-5-3-10-22-20(17)19(25)14-18/h3,5-10,13-14,25H,4,11-12H2,1-2H3,(H,23,26)
SMILES O=C(C1=CC=C(C2=CC(O)=C3N=CC=CC3=C2)C=C1)NCCCN(C)C
IUPAC Name N-(3-(dimethylamino)propyl)-4-(8-hydroxyquinolin-6-yl)benzamide
Molecular Weight (g/mol) 349.43
Formula Weight 349.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.