missing translation for 'onlineSavingsMsg'
Learn More

ML324, MedChemExpress

Product Code. 16471507
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16471507

Brand: Medchem Express HY12725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML324 is a potent JMJD2 demethylase inhibitor with antiviral activity. ML324 also exhibits inhibition for the histone demethylase KDM4B, with an IC50 of 4.9 μM. ML324 has potent anti-viral activity against both herpes simplex virus (HSV) and human cytomegalovirus (hCMV) infection via inhibition viral IE gene expression.

ML324 is a potent JMJD2 demethylase inhibitor with antiviral activity. ML324 also exhibits inhibition for the histone demethylase KDM4B, with an IC50 of 4.9 μM. ML324 has potent anti-viral activity against both herpes simplex virus (HSV) and human cytomegalovirus (hCMV) infection via inhibition viral IE gene expression.

Specifications

Chemical Name or Material ML324
Physical Form Solid
CAS 1222800-79-4
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C21H23N3O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 31.25 mg/mL (89.43 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(NCCCN(C)C)C1=CC=C(C2=CC(O)=C3N=CC=CC3=C2)C=C1
Molecular Weight (g/mol) 349.43
Formula Weight 349.43
Percent Purity 96.85%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.