missing translation for 'onlineSavingsMsg'
Learn More

ML324, MedChemExpress

Product Code. 16475956
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475956 100 mg 100mg
16471507 50 mg 50mg
16405308 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16475956 Supplier Medchem Express Supplier No. HY12725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML324 is a potent JMJD2 demethylase inhibitor with antiviral activity. ML324 also exhibits inhibition for the histone demethylase KDM4B, with an IC50 of 4.9 μM. ML324 has potent anti-viral activity against both herpes simplex virus (HSV) and human cytomegalovirus (hCMV) infection via inhibition viral IE gene expression.

ML324 is a potent JMJD2 demethylase inhibitor with antiviral activity. ML324 also exhibits inhibition for the histone demethylase KDM4B, with an IC50 of 4.9 μM. ML324 has potent anti-viral activity against both herpes simplex virus (HSV) and human cytomegalovirus (hCMV) infection via inhibition viral IE gene expression.

Specifications

Chemical Name or Material ML324
Physical Form Solid
CAS 1222800-79-4
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H23N3O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 31.25 mg/mL (89.43 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(NCCCN(C)C)C1=CC=C(C2=CC(O)=C3N=CC=CC3=C2)C=C1
Molecular Weight (g/mol) 349.43
Formula Weight 349.43
Percent Purity 96.85%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.