missing translation for 'onlineSavingsMsg'
Learn More

ML314, MedChemExpress

Product Code. 16473137
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16473137

Brand: Medchem Express HY16639

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML314 is a potent molecule agonist of NTR1 (EC50 = 1.9 μM); showed good selectivity against NTR2 and GPR35, but did not stimulate Ca2+ mobilization.

ML314 is a potent molecule agonist of NTR1 (EC50 = 1.9 μM)

Specifications

Chemical Name or Material ML314
CAS 1448895-09-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H28N4O3
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (118.91 mM; Need ultrasonic)
SMILES COC1=CC=CC=C1N2CCN(C3=C4C=C(OC)C(OC)=CC4=NC(C5CC5)=N3)CC2
Molecular Weight (g/mol) 420.5
Formula Weight 420.5
Percent Purity 99.82%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.