missing translation for 'onlineSavingsMsg'
Learn More

ML311, MedChemExpress

Product Code. 16476287
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476287 5 mg 5mg
16449976 50 mg 50mg
16444676 100 mg 100mg
16433568 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476287 Supplier Medchem Express Supplier No. HY101778

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML311 is a potent and selective inhibitor of the Mcl-1/Bim interaction.

ML311 is a potent and selective inhibitor of the Mcl-1/Bim interaction.

Specifications

Chemical Name or Material ML311
CAS 315698-17-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H24F3N3O
Quantity 5 mg
Solubility Information DMSO : 67.5 mg/mL (162.47 mM; Need ultrasonic)
SMILES OC1=C2N=CC=CC2=CC=C1C(N3CCN(CC)CC3)C4=CC=C(C(F)(F)F)C=C4
Molecular Weight (g/mol) 415.45
Formula Weight 415.45
Percent Purity 98.26%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.